Assay Details
Target Gene Details
Entrez Gene ID: | 227334 |
Gene Name: | ubiquitin specific peptidase 40 |
Gene Aliases: |
B230215L03Rik, C730029K03 |
Location: |
Chr.1:87945121-88008577 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp40 | NM_001033291.2 | 28 | 3996 | NP_001028463.1 |
| NM_001198573.1 | 32 | 4263 | NP_001185502.1 | |
| AK132295.1 | 28 | 3996 | BAE21086.1 | |
| AK153777.1 | 1 | 697 | ||
| AK165849.1 | 8 | 1232 | BAE38411.1 |
More Information
Set Membership: |
|

Genomic Map