Assay Details
Target Gene Details
Entrez Gene ID: | 114644 |
Gene Name: | solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3 |
Gene Aliases: |
NaDC-3, NaDC3, SDCT2 |
Location: |
Chr.2:165405295-165473225 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc13a3 | NM_054055.2 | 13 | 1928 | NP_473396.1 |
| AF306491.1 | 13 | 1927 | AAK49898.1 | |
| AK085326.1 | 13 | 1927 | ||
| AK087518.1 | 13 | 1929 | ||
| AK128982.1 | 13 | 1925 | ||
| AK138344.1 | 13 | 1926 | BAE23629.1 | |
| AK157881.1 | 13 | 1897 | ||
| AK165355.1 | 13 | 1931 | ||
| BC026803.1 | 13 | 1914 | AAH26803.1 |
More Information
Set Membership: |
|

Genomic Map