Assay Details
Target Gene Details
Entrez Gene ID: | 18583 |
Gene Name: | phosphodiesterase 7A |
Gene Aliases: |
AU015378, AW047537 |
Location: |
Chr.3:19223108-19311374 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde7a | NM_001122759.2 | 1 | 313 | NP_001116231.1 |
| AK154521.1 | 1 | 130 | BAE32648.1 | |
| AY007702.1 | AAG16295.1 | |||
| BC062909.1 | 1 | 312 | AAH62909.1 |
More Information
Set Membership: |
|

Genomic Map