Assay Details
Target Gene Details
Entrez Gene ID: | 12163 |
Gene Name: | bone morphogenetic protein 8a |
Gene Aliases: |
Bmp7r1, OP-2, OP2 |
Location: |
Chr.4:123312645-123343252 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bmp8a | NM_001256019.1 | 7 | 1895 | NP_001242948.1 |
| NM_007558.3 | 7 | 1856 | NP_031584.1 | |
| AK157978.1 | 7 | 1518 | BAE34297.1 | |
| BC052168.1 | 7 | 1893 | AAH52168.1 | |
| M97017.1 | 7 | 1404 | AAB01365.1 |
More Information
Set Membership: |
|

Genomic Map