Assay Details
Target Gene Details
Entrez Gene ID: | 140475 |
Gene Name: | Bartter syndrome, infantile, with sensorineural deafness (Barttin) |
Gene Aliases: |
- |
Location: |
Chr.4:106483456-106492243 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bsnd | NM_080458.2 | 3 | 560 | NP_536706.2 |
| AF391088.1 | 3 | 557 | AAL33907.1 | |
| AK052587.1 | 3 | 561 | BAC35049.1 | |
| AY373833.1 | 3 | 437 | AAQ81629.1 | |
| BC038287.1 | 3 | 611 | AAH38287.1 |

Genomic Map