Assay Details
Target Gene Details
Entrez Gene ID: | 241514 |
Gene Name: | zinc finger protein 804A |
Gene Aliases: |
C630007C17Rik, Znf804a |
Location: |
Chr.2:82052874-82259879 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp804a | NM_175513.3 | 4 | 2965 | NP_780722.2 |
AK035949.1 | 1 | 1487 | BAC29256.1 | |
BC035535.1 | 1 | 1992 | AAH35535.1 |
