Assay Details
Target Gene Details
Entrez Gene ID: | 319224 |
Gene Name: | RIKEN cDNA 6330410L21 gene |
Gene Aliases: |
- |
Location: |
Chr.3:129753758-129763882 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 6330410L21Rik | NR_040589.1 | 2 | 688 | |
| AK031828.1 | 2 | 689 |
Target Gene Details
Entrez Gene ID: | 13645 |
Gene Name: | epidermal growth factor |
Gene Aliases: |
AI790464 |
Location: |
Chr.3:129677569-129755322 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Egf | NM_010113.3 | 1 | 340 | NP_034243.2 |
| AK084927.1 | 1 | 335 | ||
| AK142666.1 | ||||
| BC017681.1 | 1 | 328 | AAH17681.1 | |
| BC060741.1 | 1 | 325 | AAH60741.1 | |
| BC092277.1 | 1 | 325 | AAH92277.1 | |
| J00380.1 | 1 | 336 | AAA37539.1 | |
| V00741.1 | 1 | 291 | CAA24115.1 |
More Information
Set Membership: |
|

Genomic Map