Assay Details
Target Gene Details
Entrez Gene ID: | 13645 |
Gene Name: | epidermal growth factor |
Gene Aliases: |
AI790464 |
Location: |
Chr.3:129677569-129755322 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Egf | NM_010113.3 | 24 | 4104 | NP_034243.2 |
| AK085707.1 | 24 | 3671 | ||
| AK088408.1 | 18 | 2568 | ||
| AK136439.1 | 19 | 2697 | BAE22978.1 | |
| AK142666.1 | 24 | 3801 | ||
| BC017681.1 | 25 | 4041 | AAH17681.1 | |
| BC060741.1 | 24 | 4089 | AAH60741.1 | |
| BC092277.1 | 24 | 4089 | AAH92277.1 | |
| J00380.1 | 24 | 4099 | AAA37539.1 | |
| V00741.1 | 24 | 4055 | CAA24115.1 |
More Information
Set Membership: |
|

Genomic Map