Assay Details
Target Gene Details
Entrez Gene ID: | 229317 |
Gene Name: | eukaryotic translation initiation factor 2A |
Gene Aliases: |
D030048D22, D3Ertd194e |
Location: |
Chr.3:58525821-58557501 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2a | NM_001005509.2 | 10 | 1520 | NP_001005509.1 |
| AK044865.1 | 4 | 1567 | ||
| AK050971.1 | 10 | 1237 | ||
| AK078519.1 | 10 | 1299 | BAC37320.1 | |
| AK145275.1 | 10 | 1476 | ||
| BC030447.1 | 9 | 1189 | ||
| BC082557.1 | 10 | 1325 | AAH82557.1 |
More Information
Set Membership: |
|

Genomic Map