Assay Details
Target Gene Details
Entrez Gene ID: | 208691 |
Gene Name: | eukaryotic translation initiation factor 5A2 |
Gene Aliases: |
9630038B20, eIF5AII |
Location: |
Chr.3:28781311-28798846 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif5a2 | NM_177586.5 | 5 | 2017 | NP_808254.1 |
| AK036127.1 | 5 | 2019 | ||
| AK052412.1 | 5 | 1961 | BAC34978.1 | |
| AK082214.1 | 5 | 1934 | BAC38441.1 | |
| AY205264.1 | 5 | 2017 | AAO18682.1 | |
| AY205265.1 | 5 | 2017 | AAO18683.1 |
More Information
Set Membership: |
|

Genomic Map