Assay Details
Target Gene Details
Entrez Gene ID: | 100039684 |
Gene Name: | RIKEN cDNA 5031434O11 gene |
Gene Aliases: |
AI481280, Gm2370 |
Location: |
Chr.3:51559757-51567117 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
5031434O11Rik | NR_033624.1 | |||
AI481280.1 | ||||
AK017144.1 | ||||
AK048747.1 | ||||
BB385113.2 | ||||
BX528204.1 |
