Assay Details
Target Gene Details
Entrez Gene ID: | 20193 |
Gene Name: | S100 calcium binding protein A1 |
Gene Aliases: |
AI266795, S100, S100a |
Location: |
Chr.3:90511034-90514330 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a1 | NM_011309.3 | 3 | 339 | NP_035439.1 |
| AF087687.1 | 3 | 306 | AAC64108.1 | |
| AK002721.1 | 3 | 280 | BAB22308.1 | |
| AK012578.1 | 3 | 339 | BAB28330.1 | |
| AK017279.1 | 3 | 275 | BAB30670.1 | |
| AK168822.1 | 3 | 275 | BAE40649.1 | |
| BC005590.1 | 3 | 411 | AAH05590.1 |
More Information
Set Membership: |
|

Genomic Map