Assay Details
Target Gene Details
Entrez Gene ID: | 17210 |
Gene Name: | myeloid cell leukemia sequence 1 |
Gene Aliases: |
AW556805, Mcl-1 |
Location: |
Chr.3:95658368-95663179 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mcl1 | NM_008562.3 | 1 | 638 | NP_032588.1 |
AK159504.1 | 1 | 550 | BAE35137.1 | |
BC003839.1 | 1 | 567 | AAH03839.1 | |
BC005427.1 | 1 | 560 | AAH05427.1 | |
BC021638.1 | 1 | 577 | AAH21638.1 | |
GU182318.1 | 2 | 364 | ||
U35623.1 | 1 | 550 | AAC31790.1 |
