Assay Details
Target Gene Details
Entrez Gene ID: | 17210 |
Gene Name: | myeloid cell leukemia sequence 1 |
Gene Aliases: |
AW556805, Mcl-1 |
Location: |
Chr.3:95658368-95663179 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mcl1 | NM_008562.3 | 3 | 2665 | NP_032588.1 |
AK144229.1 | 1 | 1215 | ||
AK148607.1 | 1 | 1205 | ||
AK159504.1 | 3 | 2577 | BAE35137.1 | |
AK160634.1 | 1 | 414 | ||
BC021638.1 | 3 | 2604 | AAH21638.1 | |
U35623.1 | 3 | 2577 | AAC31790.1 |
