Assay Details
Target Gene Details
Entrez Gene ID: | 100505386 |
Gene Name: | predicted gene, 21949 |
Gene Aliases: |
Iqcj-schip1, Schip-1-iqcj, Schip1 |
Location: |
Chr.3:67892220-68626482 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm21949 | NM_001113419.2 | 5 | 494 | NP_001106890.1 |
KJ941154.1 |
Target Gene Details
Entrez Gene ID: | 30953 |
Gene Name: | schwannomin interacting protein 1 |
Gene Aliases: |
Nf2ip, Schip-1 |
Location: |
Chr.3:68064802-68626482 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Schip1 | NM_001113420.1 | NP_001106891.1 | ||
NM_001113421.1 | 2 | 316 | NP_001106892.1 | |
NM_001282044.1 | 2 | 299 | NP_001268973.1 | |
NM_001282045.1 | 2 | 157 | NP_001268974.1 | |
AK083464.1 | 2 | 142 | ||
AK146930.1 | 2 | 273 | BAE27541.1 | |
AK168005.1 | 2 | 226 | BAE39993.1 | |
BC094348.1 | 2 | 225 | AAH94348.1 | |
BY281727.1 | 2 | 294 | ||
BY299001.1 | 2 | 287 | ||
CA750719.1 | 2 | 157 | ||
EU163407.1 | 2 | 316 | ||
EU163408.1 |
