Assay Details
Target Gene Details
Entrez Gene ID: | 100505386 |
Gene Name: | predicted gene, 21949 |
Gene Aliases: |
Iqcj-schip1, Schip-1-iqcj, Schip1 |
Location: |
Chr.3:67892220-68626482 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm21949 | NM_001113419.2 | NP_001106890.1 | ||
KJ941154.1 |
Target Gene Details
Entrez Gene ID: | 30953 |
Gene Name: | schwannomin interacting protein 1 |
Gene Aliases: |
Nf2ip, Schip-1 |
Location: |
Chr.3:68064802-68626482 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Schip1 | NM_001113420.1 | NP_001106891.1 | ||
NM_001113421.1 | NP_001106892.1 | |||
NM_001282044.1 | NP_001268973.1 | |||
NM_001282045.1 | NP_001268974.1 | |||
NM_013928.5 | 1 | 146 | NP_038956.2 | |
AF145716.1 | AAF34244.1 | |||
AK021361.1 | BAB32387.1 | |||
AK083464.1 | ||||
AK146930.1 | BAE27541.1 | |||
AK168005.1 | BAE39993.1 | |||
BC060529.1 | 1 | 80 | AAH60529.1 | |
BC094348.1 | AAH94348.1 | |||
BY243167.1 | 1 | 147 | ||
EU163407.1 | ||||
EU163408.1 | ||||
KM233716.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
