Assay Details
Target Gene Details
Entrez Gene ID: | 100505386 |
Gene Name: | predicted gene, 21949 |
Gene Aliases: |
Iqcj-schip1, Schip-1-iqcj, Schip1 |
Location: |
Chr.3:67892220-68626482 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm21949 | NM_001113419.2 | 11 | 2065 | NP_001106890.1 |
Target Gene Details
Entrez Gene ID: | 30953 |
Gene Name: | schwannomin interacting protein 1 |
Gene Aliases: |
Nf2ip, Schip-1 |
Location: |
Chr.3:68064802-68626482 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Schip1 | NM_001113420.1 | 7 | 1203 | NP_001106891.1 |
NM_001113421.1 | 8 | 1887 | NP_001106892.1 | |
NM_001282044.1 | 8 | 1870 | NP_001268973.1 | |
NM_001282045.1 | 8 | 1728 | NP_001268974.1 | |
NM_013928.5 | 7 | 1293 | NP_038956.2 | |
AF145716.1 | 7 | 1399 | AAF34244.1 | |
AK021361.1 | 7 | 1154 | BAB32387.1 | |
AK083464.1 | 8 | 1713 | ||
AK146930.1 | 8 | 1844 | BAE27541.1 | |
AK168005.1 | 8 | 1797 | BAE39993.1 | |
AW492234.1 | 1 | 331 | ||
BB440451.2 | 1 | 329 | ||
BC060529.1 | 7 | 1227 | AAH60529.1 | |
BC094348.1 | 8 | 1654 | AAH94348.1 | |
BY616163.1 | 1 | 91 | ||
BY623736.1 | 1 | 52 | ||
EU163407.1 | 8 | 1887 | ||
EU163408.1 | 7 | 1203 |
