Assay Details
Target Gene Details
Entrez Gene ID: | 21420 |
Gene Name: | transcription factor AP-2, gamma |
Gene Aliases: |
AA409384, AP2gamma, Ap-2.2, Stra2, Tcfap2c |
Location: |
Chr.2:172545678-172558622 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tfap2c | NM_001159696.1 | 2 | 393 | NP_001153168.1 |
| NM_009335.2 | 2 | 452 | NP_033361.2 | |
| AK050973.1 | 2 | 370 | ||
| AK141211.1 | 2 | 354 | BAE24590.1 | |
| AK145166.1 | 2 | 402 | BAE26271.1 | |
| AK145603.1 | 2 | 347 | BAE26536.1 | |
| BC003778.1 | 2 | 326 | AAH03778.1 | |
| CJ080187.1 | ||||
| KJ463836.1 | 2 | 248 | ||
| X94694.1 | 2 | 431 | CAA64357.1 |
More Information
Set Membership: |
|

Genomic Map