Assay Details
Target Gene Details
Entrez Gene ID: | 228880 |
Gene Name: | zinc finger, MYND-type containing 8 |
Gene Aliases: |
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7 |
Location: |
Chr.2:165784152-165898984 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmynd8 | NM_001252584.2 | NP_001239513.1 | ||
| NM_001252585.1 | NP_001239514.1 | |||
| NM_001281926.1 | NP_001268855.1 | |||
| NM_001291158.1 | 16 | 4715 | NP_001278087.1 | |
| NM_027230.5 | 16 | 2793 | NP_081506.3 | |
| AB093284.2 | 16 | 4707 | BAC41468.4 | |
| AK014397.1 | 3 | 850 | BAE43241.1 | |
| AK132427.1 | BAE21162.1 | |||
| AK147620.1 | BAE28029.1 | |||
| AK155864.1 | BAE33468.1 | |||
| BC041797.1 | 5 | 1521 | AAH41797.1 | |
| BC048186.1 | 16 | 2600 | AAH48186.3 |

Genomic Map