Assay Details
Target Gene Details
Entrez Gene ID: | 228880 |
Gene Name: | zinc finger, MYND-type containing 8 |
Gene Aliases: |
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7 |
Location: |
Chr.2:165784152-165898984 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmynd8 | NM_001252584.2 | 23 | 4385 | NP_001239513.1 |
| NM_001252585.1 | 22 | 4277 | NP_001239514.1 | |
| NM_001281926.1 | 23 | 4284 | NP_001268855.1 | |
| NM_001291158.1 | 24 | 6550 | NP_001278087.1 | |
| NM_027230.5 | 24 | 4628 | NP_081506.3 | |
| AK008125.1 | 1 | 415 | BAB25478.1 | |
| AK139219.1 | 1 | 751 | BAE23921.1 | |
| AK147620.1 | 23 | 4285 | BAE28029.1 | |
| AK155864.1 | 23 | 4190 | BAE33468.1 |

Genomic Map