Assay Details
Target Gene Details
Entrez Gene ID: | 329628 |
Gene Name: | FAT tumor suppressor homolog 4 (Drosophila) |
Gene Aliases: |
6030410K14Rik, 9430004M15 |
Location: |
Chr.3:38886487-39011987 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fat4 | NM_183221.3 | 16 | 12962 | NP_899044.3 |
| BC079887.1 | 8 | 4056 | AAH79887.1 | |
| DQ286572.1 | 16 | 12962 | ABB88946.1 |
More Information
Set Membership: |
|

Genomic Map