Assay Details
Target Gene Details
Entrez Gene ID: | 100039684 |
Gene Name: | RIKEN cDNA 5031434O11 gene |
Gene Aliases: |
AI481280, Gm2370 |
Location: |
Chr.3:51559757-51567117 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5031434O11Rik | NR_033624.1 | |||
| AK048747.1 | ||||
| BX528204.1 |
Target Gene Details
Entrez Gene ID: | 73251 |
Gene Name: | SET domain containing (lysine methyltransferase) 7 |
Gene Aliases: |
1600028F23Rik, H3K4MT, KMT7, Set7, Set7/9, mKIAA1717 |
Location: |
Chr.3:51515318-51560823 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setd7 | NM_080793.5 | 1 | 106 | NP_542983.3 |
| AK048924.1 | BAC33493.1 | |||
| AK147316.1 | 1 | 162 | ||
| AK147413.1 | BAE27897.1 | |||
| AK147422.1 | 1 | 107 | BAE27903.1 | |
| AK147667.1 | 1 | 140 | BAE28059.1 | |
| AK170161.1 | BAE41607.1 | |||
| BC050190.1 | AAH50190.1 |
More Information
Set Membership: |
|

Genomic Map