Assay Details
Target Gene Details
Entrez Gene ID: | 20568 |
Gene Name: | secretory leukocyte peptidase inhibitor |
Gene Aliases: |
- |
Location: |
Chr.2:164354070-164356507 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slpi | NM_011414.3 | 1 | 51 | NP_035544.1 |
| BC028509.1 | 1 | 51 | AAH28509.1 |
More Information
Set Membership: |
|

Genomic Map