Assay Details
Target Gene Details
Entrez Gene ID: | 66350 |
Gene Name: | phospholipase A2, group XIIA |
Gene Aliases: |
2310004B05Rik, GXII, GXII-1, GXII-1-PLA2, Pla2g12, Rossy, mGXII-1 |
Location: |
Chr.3:129878606-129895825 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g12a | NM_001286948.1 | 4 | 1465 | NP_001273877.1 |
| NM_023196.4 | 4 | 1396 | NP_075685.2 | |
| NM_183423.3 | 4 | 1179 | NP_904359.1 | |
| NR_104611.1 | 5 | 1554 | ||
| AK009133.1 | 4 | 1398 | BAB26094.1 | |
| AK163693.1 | 4 | 1310 | BAE37460.1 | |
| AK195519.1 | 1 | 49 | ||
| AY007381.1 | 4 | 1369 | AAG23336.1 | |
| BB505724.1 | 1 | 97 | ||
| BC051117.1 | 4 | 1396 | AAH51117.1 |
More Information
Set Membership: |
|

Genomic Map