Assay Details
Target Gene Details
Entrez Gene ID: | 27371 |
Gene Name: | SH2 domain containing 2A |
Gene Aliases: |
Lad, Ribp, Tsad |
Location: |
Chr.3:87846755-87855722 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh2d2a | NM_001025571.2 | 7 | 2906 | NP_001020742.1 |
| NM_021309.3 | 7 | 2930 | NP_067284.1 | |
| AK037489.1 | 7 | 2628 | BAC29819.1 |
More Information
Set Membership: |
|

Genomic Map