Assay Details
Target Gene Details
Entrez Gene ID: | 72535 |
Gene Name: | aldehyde dehydrogenase 1 family, member B1 |
Gene Aliases: |
2700007F14Rik |
Location: |
Chr.4:45799022-45804608 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh1b1 | NM_028270.4 | 2 | 867 | NP_082546.1 |
| AK012213.1 | 2 | 762 | BAB28101.1 | |
| AK088396.1 | 2 | 841 | BAC40326.1 | |
| AK150992.1 | 2 | 769 | BAE30016.1 | |
| AK151349.1 | 2 | 769 | BAE30325.1 | |
| AK151364.1 | 2 | 769 | BAE30339.1 | |
| AK153416.1 | 2 | 769 | BAE31976.1 | |
| BC020001.1 | 2 | 771 | AAH20001.1 | |
| BC086768.1 | 2 | 826 | AAH86768.1 |
More Information
Set Membership: |
|

Genomic Map