Assay Details
Target Gene Details
Entrez Gene ID: | 21969 |
Gene Name: | topoisomerase (DNA) I |
Gene Aliases: |
AI467334, D130064I21Rik, Top-1 |
Location: |
Chr.2:160645897-160722764 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top1 | NM_009408.2 | 1 | 125 | NP_033434.2 |
| AK083930.1 | 1 | 42 | ||
| AK145538.1 | 1 | 135 | BAE26493.1 | |
| AK160590.1 | BAE35897.1 | |||
| AK164061.1 | 1 | 44 | BAE37608.1 | |
| AK166844.1 | 1 | 42 | ||
| CF733054.1 | 1 | 125 |
More Information
Set Membership: |
|

Genomic Map