Assay Details
Target Gene Details
Entrez Gene ID: | 229499 |
Gene Name: | Fc receptor-like 1 |
Gene Aliases: |
A230020G22Rik, Bxmas1, Bxmh1, Bxmh1b, Fcrh1, Fcrh1l, Fcrh1s, Ifgp1 |
Location: |
Chr.3:87376333-87402934 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcrl1 | NM_001136236.1 | 3 | 204 | NP_001129708.1 |
| NM_153090.2 | 4 | 264 | NP_694730.2 | |
| NM_178165.4 | 3 | 204 | NP_835459.1 | |
| AF329485.1 | 3 | 131 | AAM11542.1 | |
| AK038494.1 | 3 | 144 | BAC30017.1 | |
| AK041750.1 | 4 | 266 | ||
| AK156528.1 | 3 | 144 | ||
| AK156985.1 | 3 | 144 | BAE33923.1 | |
| AY158088.1 | 3 | 124 | AAO20871.1 | |
| AY158089.1 | 3 | 124 | AAO20872.1 | |
| AY506554.1 | 3 | 137 | AAS91574.1 | |
| AY506555.1 | 3 | 137 | AAS91575.1 | |
| BC055830.1 | 3 | 131 | AAH55830.1 |
More Information
Set Membership: |
|

Genomic Map