Assay Details
Target Gene Details
Entrez Gene ID: | 18805 |
Gene Name: | phospholipase D1 |
Gene Aliases: |
AA536939, C85393, Pld1a, Pld1b |
Location: |
Chr.3:27938680-28133362 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pld1 | NM_001164056.1 | 27 | 4564 | NP_001157528.1 |
| NM_008875.4 | 26 | 4509 | NP_032901.2 | |
| AK135690.1 | 2 | 1486 | ||
| AK136111.1 | 22 | 3816 | BAE22827.1 | |
| BC068144.1 | 26 | 4366 | AAH68144.1 |
More Information
Set Membership: |
|

Genomic Map