Assay Details
Target Gene Details
Entrez Gene ID: | 140629 |
Gene Name: | U box domain containing 5 |
Gene Aliases: |
1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5 |
Location: |
Chr.2:130589996-130630038 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubox5 | NM_001255993.1 | 5 | 1506 | NP_001242922.1 |
NM_001255994.1 | 4 | 1359 | NP_001242923.1 | |
NM_080562.5 | 4 | 1364 | NP_542129.2 | |
AF360997.1 | 4 | 1294 | AAK51467.1 | |
AK034114.1 | 4 | 1313 | BAC28590.1 | |
AK049276.1 | 3 | 1130 | BAC33652.1 | |
AK158052.1 | 5 | 1464 | BAE34338.1 | |
AK170100.1 | 4 | 1335 | ||
AK171070.1 | 5 | 1473 | ||
AK177084.1 | ||||
BC025068.1 | 3 | 1226 | AAH25068.1 |
