Assay Details
Target Gene Details
Entrez Gene ID: | 14173 |
Gene Name: | fibroblast growth factor 2 |
Gene Aliases: |
Fgf-2, Fgfb, bFGF |
Location: |
Chr.3:37348469-37405538 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf2 | NM_008006.2 | 1 | 59 | NP_032032.1 |
| BY730691.1 | 1 | 37 | ||
| CJ098597.1 | 1 | 62 |
More Information
Set Membership: |
|

Genomic Map