Assay Details
Target Gene Details
Entrez Gene ID: | 69893 |
Gene Name: | cytochrome c oxidase assembly factor 7 |
Gene Aliases: |
2010305A19Rik, AI451324, D4Ertd796e, Selrc1 |
Location: |
Chr.4:108328152-108340718 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coa7 | NM_027250.4 | 3 | 711 | NP_081526.1 |
| AK008518.1 | 3 | 702 | ||
| AK044999.1 | 3 | 712 | BAC32178.1 | |
| AK081388.1 | 3 | 724 | BAC38209.1 | |
| AK168344.1 | 3 | 691 | BAE40281.1 | |
| BC012391.1 | 3 | 709 | AAH12391.1 |

Genomic Map