Assay Details
Target Gene Details
Entrez Gene ID: | 74270 |
Gene Name: | ubiquitin specific peptidase 20 |
Gene Aliases: |
1700055M05Rik, AI467231, Vdu2 |
Location: |
Chr.2:30982279-31022655 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp20 | NM_028846.5 | 25 | 3581 | NP_083122.1 |
| AF449715.1 | 25 | 3532 | AAN15803.1 | |
| AK047242.1 | 25 | 3500 | ||
| AK054279.1 | 25 | 3524 | BAC35715.1 | |
| AK087412.1 | 25 | 3493 | ||
| AK163663.1 | 25 | 3491 | BAE37447.1 | |
| AK173083.1 | 25 | 3483 | BAD32361.1 | |
| BC019735.1 | 11 | 1781 | AAH19735.1 |
More Information
Set Membership: |
|

Genomic Map