Assay Details
Target Gene Details
Entrez Gene ID: | 75083 |
Gene Name: | ubiquitin specific peptidase 50 |
Gene Aliases: |
1700086G18Rik, 4930511O11Rik |
Location: |
Chr.2:126761050-126783460 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp50 | NM_029163.3 | 7 | 1201 | NP_083439.2 |
| AK015766.1 | 7 | 1186 | BAB29966.1 | |
| AW539155.2 | ||||
| BC061020.1 | 7 | 1178 | AAH61020.1 |
More Information
Set Membership: |
|

Genomic Map