Assay Details
Target Gene Details
Entrez Gene ID: | 242248 |
Gene Name: | B cell scaffold protein with ankyrin repeats 1 |
Gene Aliases: |
A530094C12Rik, AI451642, AVIEF, BANK |
Location: |
Chr.3:136053363-136326141 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bank1 | NM_001033350.2 | 17 | 2993 | NP_001028522.2 |
AA175345.1 | ||||
AA178225.1 | ||||
AK148665.1 | 6 | 788 | BAE28636.1 | |
AK156201.1 | 17 | 3004 | BAE33622.1 | |
AY178734.1 | 17 | 3013 | AAO13613.1 | |
BC116328.1 | 7 | 1076 | ||
EU051378.1 | 16 | 2594 |
