Assay Details
Target Gene Details
Entrez Gene ID: | 242248 |
Gene Name: | B cell scaffold protein with ankyrin repeats 1 |
Gene Aliases: |
A530094C12Rik, AI451642, AVIEF, BANK |
Location: |
Chr.3:136053363-136326141 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bank1 | NM_001033350.2 | 2 | 446 | NP_001028522.2 |
AK041242.1 | 2 | 407 | BAC30875.1 | |
AK156201.1 | 2 | 456 | BAE33622.1 | |
AY178734.1 | 2 | 466 | AAO13613.1 | |
BC026932.1 | 2 | 393 | ||
BC116328.1 | ||||
BY107051.1 | ||||
EU051378.1 |
