Assay Details
Target Gene Details
Entrez Gene ID: | 229503 |
Gene Name: | ribosomal RNA adenine dimethylase domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.3:87922601-87930588 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrnad1 | NM_153562.4 | 1 | 201 | NP_705790.3 |
| AK030684.1 | 1 | 153 | ||
| AK035389.1 | 1 | 153 | BAC29055.1 | |
| AK082223.1 | 1 | 133 | ||
| AK148128.1 | 1 | 162 | ||
| AK165264.1 | 1 | 143 | ||
| AK172049.1 | ||||
| BB871494.1 | 1 | 206 | ||
| BY035934.1 | 1 | 151 |
More Information
Set Membership: |
|

Genomic Map