Assay Details
Target Gene Details
Entrez Gene ID: | 19142 |
Gene Name: | protease, serine 12 neurotrypsin (motopsin) |
Gene Aliases: |
Bssp-3, motopsin |
Location: |
Chr.3:123446913-123506602 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss12 | NM_008939.2 | 1 | 245 | NP_032965.1 |
AK132633.1 | 1 | 32 | ||
BC031429.1 | 1 | 147 | AAH31429.1 | |
D89871.1 | 1 | 245 | BAA23986.1 | |
Y13192.1 | CAA73646.1 |
