Assay Details
Target Gene Details
Entrez Gene ID: | 75526 |
Gene Name: | epididymal peptidase inhibitor |
Gene Aliases: |
1700024E17Rik, Spinlw1 |
Location: |
Chr.2:164588343-164593571 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eppin | NM_029325.2 | 4 | 610 | NP_083601.1 |
| AF346413.2 | 4 | 633 | AAK31335.1 | |
| AK006296.1 | 4 | 612 | BAB24514.1 | |
| BC048637.1 | 4 | 633 | AAH48637.1 |

Genomic Map