Assay Details
Target Gene Details
Entrez Gene ID: | 72007 |
Gene Name: | fibronectin type III domain containing 3B |
Gene Aliases: |
1600019O04Rik, AW550168, Fad104, mKIAA4164 |
Location: |
Chr.3:27416161-27711662 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fndc3b | NM_173182.2 | 6 | 873 | NP_775274.2 |
| AB098596.1 | 6 | 811 | BAC53726.1 | |
| AK045099.1 | 6 | 968 | BAC32222.1 | |
| AK147346.1 | 7 | 884 | ||
| AK220297.1 | 6 | 873 | BAD90222.1 | |
| BC067389.1 | 6 | 879 | AAH67389.1 | |
| DQ192037.1 | 5 | 779 | ABA82150.1 |
More Information
Set Membership: |
|

Genomic Map