Assay Details
Target Gene Details
Entrez Gene ID: | 67547 |
Gene Name: | solute carrier family 39 (metal ion transporter), member 8 |
Gene Aliases: |
4933419D20Rik, AA986696, BIGM103, ZIP-8, Zip8 |
Location: |
Chr.3:135816883-135888572 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a8 | NM_001135149.1 | NP_001128621.1 | ||
| NM_001135150.1 | 1 | 52 | NP_001128622.1 | |
| NM_026228.5 | NP_080504.3 | |||
| AK014895.1 | BAB29610.1 | |||
| AK016853.1 | BAB30465.1 | |||
| AK028215.1 | ||||
| AK085740.1 | BAC39526.1 | |||
| BY730883.1 | ||||
| CN528304.1 | 1 | 52 |
More Information
Set Membership: |
|

Genomic Map