Assay Details
Target Gene Details
Entrez Gene ID: | 241919 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 14 |
Gene Aliases: |
A930013N06, BC061928 |
Location: |
Chr.3:31202854-31310574 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a14 | NM_172861.3 | 8 | 4488 | NP_766449.1 |
| AK173217.1 | 8 | 4464 | BAD32495.1 | |
| BC061928.1 | 8 | 4464 | AAH61928.1 |
More Information
Set Membership: |
|

Genomic Map