Assay Details
Target Gene Details
Entrez Gene ID: | 277343 |
Gene Name: | WAP four-disulfide core domain 8 |
Gene Aliases: |
Gm706 |
Location: |
Chr.2:164596458-164613626 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wfdc8 | NM_001080550.2 | 8 | 1705 | NP_001074019.1 |
NM_001276431.1 | NP_001263360.1 | |||
NM_001276432.1 | 7 | 2391 | NP_001263361.1 | |
AK136599.1 | 7 | 2392 | ||
AY669506.1 | AAU89496.1 | |||
AY757956.1 | 8 | 1705 | AAU93641.1 |
