Assay Details
Target Gene Details
Entrez Gene ID: | 12167 |
Gene Name: | bone morphogenetic protein receptor, type 1B |
Gene Aliases: |
AI385617, ALK-6, AV355320, Acvrlk6, Alk6, BMPR-1B, BMPR-IB, CFK-43a, SKR6 |
Location: |
Chr.3:141837136-142169590 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bmpr1b | NM_001277216.1 | NP_001264145.1 | ||
| NM_001277217.1 | NP_001264146.1 | |||
| NM_007560.4 | 1 | 177 | NP_031586.1 | |
| AK086130.1 | BAC39617.1 | |||
| BC065143.1 | 1 | 164 | AAH65143.1 | |
| CJ062780.1 | ||||
| CJ133683.1 | 1 | 178 | ||
| Z23143.1 | CAA80674.1 |
More Information
Set Membership: |
|

Genomic Map