Assay Details
Target Gene Details
Entrez Gene ID: | 108058 |
Gene Name: | calcium/calmodulin-dependent protein kinase II, delta |
Gene Aliases: |
2810011D23Rik, 8030469K03Rik, CaMK II, [d]-CaMKII |
Location: |
Chr.3:126596358-126846326 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2d | NM_001025438.2 | 1 | 195 | NP_001020609.1 |
| NM_001025439.2 | 1 | 195 | NP_001020610.1 | |
| NM_001293663.1 | 1 | 195 | NP_001280592.1 | |
| NM_001293664.1 | 1 | 195 | NP_001280593.1 | |
| NM_001293665.1 | 1 | 195 | NP_001280594.1 | |
| NM_001293666.1 | 1 | 195 | NP_001280595.1 | |
| NM_023813.4 | 1 | 195 | NP_076302.1 | |
| AK032524.1 | 1 | 142 | BAC27910.1 | |
| AK042722.1 | 1 | 137 | ||
| AK044872.1 | 1 | 137 | ||
| AK142435.1 | 1 | 138 | BAE25062.1 | |
| AK145106.1 | 1 | 126 | ||
| AK157736.1 | 1 | 137 | ||
| AK220516.1 | 1 | 195 | BAD90304.1 | |
| BB855224.1 | 1 | 174 |
More Information
Set Membership: |
|

Genomic Map