Assay Details
Target Gene Details
Entrez Gene ID: | 14173 |
Gene Name: | fibroblast growth factor 2 |
Gene Aliases: |
Fgf-2, Fgfb, bFGF |
Location: |
Chr.3:37348469-37405538 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf2 |
Target Gene Details
Entrez Gene ID: | 229228 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 6 |
Gene Aliases: |
Asfgf2b |
Location: |
Chr.3:37404982-37420186 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt6 | NM_001291044.1 | 5 | 750 | NP_001277973.1 |
| NM_153561.3 | 5 | 747 | NP_705789.1 | |
| AF453427.1 | 5 | 859 | AAN76847.1 | |
| AF453428.1 | 5 | 736 | AAN76848.1 | |
| AK031663.1 | ||||
| AK145299.1 | 5 | 742 | BAE26354.1 | |
| BC027267.1 | 5 | 741 | AAH27267.1 | |
| BC146000.1 | 5 | 744 |
More Information
Set Membership: |
|

Genomic Map