Assay Details
Target Gene Details
Entrez Gene ID: | 76897 |
Gene Name: | RALY RNA binding protein-like |
Gene Aliases: |
0710005M24Rik |
Location: |
Chr.3:13471655-14182287 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralyl | NM_001163328.1 | 9 | 2206 | NP_001156800.1 |
| NM_001163329.1 | 8 | 1799 | NP_001156801.1 | |
| NM_001163330.1 | 7 | 1626 | NP_001156802.1 | |
| NM_178631.4 | 10 | 2299 | NP_848746.1 | |
| AK032086.1 | 9 | 2139 | BAC27689.1 |

Genomic Map