Assay Details
Target Gene Details
Entrez Gene ID: | 22329 |
Gene Name: | vascular cell adhesion molecule 1 |
Gene Aliases: |
CD106, Vcam-1 |
Location: |
Chr.3:116110020-116129688 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vcam1 | NM_011693.3 | 9 | 2721 | NP_035823.3 |
AK078081.1 | 2 | 520 | ||
AK143401.1 | 9 | 2491 | BAE25364.1 | |
AK162954.1 | 9 | 2491 | BAE37132.1 | |
AK169722.1 | 5 | 1435 | ||
BC011159.1 | 9 | 2463 | AAH11159.2 | |
BC029823.1 | 9 | 2721 | AAH29823.1 | |
M84487.1 | 9 | 2457 | AAA40545.1 | |
X67783.1 | 9 | 2415 | CAA47989.1 |
