Assay Details
Target Gene Details
Entrez Gene ID: | 329650 |
Gene Name: | mediator complex subunit 12-like |
Gene Aliases: |
A130035F20, A630079M02, AU044581, TRALPUSH |
Location: |
Chr.3:59005595-59318416 on Build GRCm38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med12l | NM_177855.3 | NP_808523.2 | ||
| AK047743.1 | BAC33144.1 | |||
| BP757618.1 |
Target Gene Details
Entrez Gene ID: | 74191 |
Gene Name: | purinergic receptor P2Y, G-protein coupled 13 |
Gene Aliases: |
2010001L06Rik, GPCR1, GPR94, Gpr86, P2Y13, P2yr13, SP174 |
Location: |
Chr.3:59207892-59210882 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2ry13 | NM_028808.3 | 2 | 1702 | NP_083084.2 |
| AK008013.2 | 2 | 1703 | BAB25409.1 | |
| AK038560.1 | 2 | 1703 | BAC30043.1 | |
| AK083264.1 | 2 | 1703 | BAC38835.1 |
More Information
Set Membership: |
|

Genomic Map