Assay Details
Target Gene Details
Entrez Gene ID: | 20754 |
Gene Name: | small proline-rich protein 1B |
Gene Aliases: |
- |
Location: |
Chr.3:92436809-92438779 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sprr1b | NM_009265.3 | 2 | 183 | NP_033291.1 |
| AK003398.1 | 2 | 184 | BAB22763.1 | |
| AK009752.1 | 2 | 184 | BAB26478.1 | |
| BC119288.1 | 2 | 176 | ||
| BC119290.1 | 2 | 176 | ||
| X91825.1 | 2 | 167 | CAA62934.1 |

Genomic Map